The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_057135.1 | NP_476476.1 | 7 - 8 | 1023 | 67 |
XM_006254192.2 | XP_006254254.1 | 7 - 8 | 1612 | 67 | |
XM_006254193.2 | XP_006254255.1 | 7 - 8 | 1612 | 67 | |
XM_006254195.2 | XP_006254257.1 | 7 - 8 | 1612 | 67 | |
XM_006254196.2 | XP_006254258.1 | 7 - 8 | 1612 | 67 | |
XM_006254197.2 | XP_006254259.1 | 7 - 8 | 1611 | 67 | |
XM_006254198.2 | XP_006254260.1 | 7 - 8 | 1611 | 67 | |
XM_008771850.1 | XP_008770072.1 | 7 - 8 | 1612 | 67 | |
XM_008771851.1 | XP_008770073.1 | 7 - 8 | 639 | 67 | |
GenBank mRNA | AB006710.1 | - | 6 - 7 | 438 | 67 |
D87240.1 | - | 7 - 8 | 1023 | 67 | |
D87241.1 | - | 7 - 8 | 1023 | 67 | |
D87242.1 | - | 7 - 8 | 1023 | 67 | |
D87243.1 | - | 7 - 8 | 1023 | 67 | |
D87244.1 | - | 7 - 8 | 1023 | 67 | |
D87245.1 | - | 7 - 8 | 1023 | 67 | |
D87246.1 | - | 7 - 8 | 1023 | 67 | |
D87247.1 | - | 7 - 8 | 1023 | 67 | |
EU034674.1 | - | 7 - 8 | 618 | 67 |