Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lphn3
Entrez Gene ID
170641
Gene Name
latrophilin 3
Gene Aliases
Cirl3, Lec3
Chromosome Location
Chr.14: 28362176 - 28856419 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130822.1 NP_570835.1 14 - 15 2726 98
XM_008769968.1 XP_008768190.1 14 - 15 2830 98
XM_008769969.1 XP_008768191.1 14 - 15 2815 98
XM_008769970.1 XP_008768192.1 14 - 15 2830 98
XM_008769971.1 XP_008768193.1 14 - 15 2830 98
XM_008769972.1 XP_008768194.1 13 - 14 2791 98
XM_008769973.1 XP_008768195.1 14 - 15 2830 98
XM_008769974.1 XP_008768196.1 14 - 15 2830 98
XM_008769975.1 XP_008768197.1 14 - 15 2830 98
XM_008769976.1 XP_008768198.1 14 - 15 2830 98
XM_008769977.1 XP_008768199.1 13 - 14 2626 98
XM_008769978.1 XP_008768200.1 13 - 14 2791 98
XM_008769979.1 XP_008768201.1 13 - 14 2626 98
XM_008769980.1 XP_008768202.1 14 - 15 2830 98
XM_008769981.1 XP_008768203.1 14 - 15 2830 98
XM_008769982.1 XP_008768204.1 14 - 15 2829 98
XM_008769983.1 XP_008768205.1 14 - 15 2829 98
XM_008769984.1 XP_008768206.1 14 - 15 2829 98
XM_008769985.1 XP_008768207.1 13 - 14 2625 98
XM_008769986.1 XP_008768208.1 13 - 14 2625 98
GenBank mRNA AF063103.1 - 14 - 15 2726 98
AF081154.1 - 13 - 14 2547 98
AF081155.1 - 13 - 14 2547 98
AF081156.1 - 13 - 14 2547 98
AF081157.1 - 14 - 15 2751 98
AF081158.1 - 14 - 15 2751 98
AF081159.1 - 14 - 15 2751 98

View Full Product Details