Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gcg
Entrez Gene ID
24952
Gene Name
glucagon
Gene Aliases
GLP-1
Chromosome Location
Chr.3: 48442635 - 48451650 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012707.2 NP_036839.2 3 - 4 320 73
GenBank mRNA BP479145.1 - 3 - 4 320 73
BP480555.1 - 3 - 4 295 73

View Full Product Details