Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tnfrsf1a
Entrez Gene ID
25625
Gene Name
tumor necrosis factor receptor superfamily, member 1a
Gene Aliases
TNFR-1, Tnfr1
Chromosome Location
Chr.4: 157864916 - 157877634 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013091.1 NP_037223.1 4 - 5 715 94
XM_006237334.2 XP_006237396.1 3 - 4 950 94
GenBank mRNA AF329976.1 - 4 - 5 714 94
AF329977.1 - 4 - 5 714 94
AF329978.1 - 4 - 5 714 94
AF329979.1 - 4 - 5 714 94
AF329980.1 - 4 - 5 714 94
AF329981.1 - 4 - 5 714 94
BC086413.1 - 4 - 5 735 94
M63122.1 - 4 - 5 715 94

View Full Product Details