Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pnlip
Entrez Gene ID
25702
Gene Name
pancreatic lipase
Gene Aliases
PANLI
Chromosome Location
Chr.1: 279798187 - 279811372 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013161.2 NP_037293.1 9 - 10 959 72
GenBank mRNA D88534.1 - 8 - 9 931 72
D88535.1 - 8 - 9 931 72
M58369.1 - 9 - 10 960 72

View Full Product Details