The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_134369.1 | NP_599196.1 | 7 - 8 | 1170 | 63 |
XM_006228558.2 | XP_006228620.1 | 7 - 8 | 1129 | 63 | |
GenBank mRNA | AF368269.1 | - | 7 - 8 | 1170 | 63 |