Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Cyp4f5
- Entrez Gene ID
-
286905
- Gene Name
- cytochrome P450, family 4, subfamily f, polypeptide 5
- Gene Aliases
-
-
- Chromosome Location
-
Chr.7: 14559806 - 14574345 on Build Rnor_6.0
- Gene Symbol
- Cyp4f37
- Entrez Gene ID
-
691312
- Gene Name
- cytochrome P450, family 4, subfamily f, polypeptide 37
- Gene Aliases
-
-
- Chromosome Location
-
Chr.7: 14589520 - 14614869 on Build Rnor_6.0
- Gene Symbol
- LOC100912471
- Entrez Gene ID
-
100912471
- Gene Name
- cytochrome P450 4F5-like
- Gene Aliases
-
-
- Chromosome Location
-
Chr.7: 14760053 - 14830055 on Build Rnor_6.0
- Gene Symbol
- LOC687345
- Entrez Gene ID
-
687345
- Gene Name
- similar to cytochrome P450 4F1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.7: 14816009 - 14820162 on Build Rnor_6.0
- Gene Symbol
- LOC691320
- Entrez Gene ID
-
691320
- Gene Name
- similar to cytochrome P450 4F1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.7: 13881303 - 13889392 on Build Rnor_6.0
View Full Product Details