The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017092.1 | NP_058788.1 | 18 - 19 | 2274 | 70 |
GenBank mRNA | D37880.1 | - | 18 - 19 | 2274 | 70 |