Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pkn1
Entrez Gene ID
29355
Gene Name
protein kinase N1
Gene Aliases
PAK-1, Prkcl1
Chromosome Location
Chr.19: 24803524 - 24831500 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017175.2 NP_058871.2 21 - 22 2809 105
XM_006255245.2 XP_006255307.1 21 - 22 3849 105
GenBank mRNA BC061836.1 - 21 - 22 2710 105
D26180.1 - 21 - 22 2760 105
EV774518.1 - 4 - 5 429 105
FQ222217.1 - 21 - 22 2809 105
L35634.1 - 21 - 22 2652 105

View Full Product Details