Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
H2afy
Entrez Gene ID
29384
Gene Name
H2A histone family, member Y
Gene Aliases
-
Chromosome Location
Chr.17: 9282106 - 9344698 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017182.2 NP_058878.1 8 - 9 1132 60
XM_006253573.2 XP_006253635.1 8 - 9 1170 60
XM_006253574.2 XP_006253636.1 8 - 9 1260 60
XM_006253575.2 XP_006253637.1 8 - 9 1161 60
XM_006253576.2 XP_006253638.1 8 - 9 1158 60
GenBank mRNA BC089093.1 - 8 - 9 1132 60
M99065.2 - 7 - 8 973 60
U79139.1 - 8 - 9 1077 60

View Full Product Details