The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017223.2 | NP_058919.1 | 9 - 10 | 2127 | 57 |
XM_008771351.1 | XP_008769573.1 | 9 - 10 | 3134 | 57 | |
XM_008771352.1 | XP_008769574.1 | 9 - 10 | 2774 | 57 | |
GenBank mRNA | BC070908.1 | - | 9 - 10 | 2127 | 57 |
L19931.1 | - | 8 - 9 | 1983 | 57 |