Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Eci1
Entrez Gene ID
29740
Gene Name
enoyl-CoA delta isomerase 1
Gene Aliases
Dci
Chromosome Location
Chr.10: 13797573 - 13810918 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017306.4 NP_059002.2 3 - 4 260 56
GenBank mRNA BC078705.1 - 3 - 4 260 56
FQ218351.1 - 3 - 4 261 56
M61112.1 - 3 - 4 249 56
X61184.1 - 3 - 4 271 56

View Full Product Details