Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Resp18
Entrez Gene ID
50561
Gene Name
regulated endocrine-specific protein 18
Gene Aliases
-
Chromosome Location
Chr.9: 82470794 - 82477136 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019278.1 NP_062151.1 4 - 5 478 74
GenBank mRNA BC058147.1 - 4 - 5 474 74
L25633.1 - 4 - 5 478 74

View Full Product Details