Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Akp3
Entrez Gene ID
64621
Gene Name
alkaline phosphatase 3, intestine, not Mn requiring
Gene Aliases
Alpi2, IAP-2, IAP-II
Chromosome Location
Chr.9: 94228960 - 94232001 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022680.1 NP_073171.1 1 - 2 68 76

View Full Product Details