Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mfge8
Entrez Gene ID
25277
Gene Name
milk fat globule-EGF factor 8 protein
Gene Aliases
AGS, MFGME8, MFGMP-E8, OAcGD3S
Chromosome Location
Chr.1: 140845478 - 140860882 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040186.2 NP_001035276.1 6 - 7 916 73
NM_012811.3 NP_036943.1 5 - 6 748 73
GenBank mRNA BC085754.1 - 5 - 6 701 73
D84068.1 - 5 - 6 708 73
DQ455823.1 - 6 - 7 869 73

View Full Product Details