The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138519.2 | NP_612528.2 | 7 - 8 | 970 | 90 |
XM_006230009.2 | XP_006230071.1 | 7 - 8 | 978 | 90 | |
GenBank mRNA | AF245040.1 | - | 6 - 7 | 836 | 90 |
BC085702.1 | - | 6 - 7 | 830 | 90 | |
BC160821.1 | - | 7 - 8 | 970 | 90 |