Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Trim50
Entrez Gene ID
288596
Gene Name
tripartite motif-containing 50
Gene Aliases
-
Chromosome Location
Chr.12: 24348426 - 24365470 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181080.1 NP_851594.1 2 - 3 501 70
XM_006249128.2 XP_006249190.1 3 - 4 823 70
XM_006249129.2 XP_006249191.1 3 - 4 823 70
GenBank mRNA AY081950.1 - 2 - 3 501 70
BC085684.1 - 3 - 4 661 70

View Full Product Details