The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_181091.2 | NP_851605.1 | 3 - 4 | 205 | 119 |
XM_006228606.2 | XP_006228668.1 | 4 - 5 | 503 | 119 | |
XM_008759100.1 | XP_008757322.1 | 4 - 5 | 318 | 119 | |
XM_008759101.1 | XP_008757323.1 | 3 - 4 | 247 | 119 | |
XM_008759102.1 | XP_008757324.1 | 3 - 4 | 195 | 119 | |
GenBank mRNA | AB007364.1 | - | 3 - 4 | 205 | 119 |