Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Klk1c7
- Entrez Gene ID
-
24523
- Gene Name
- kallikrein 1-related peptidase C7
- Gene Aliases
-
KALA, Kal, Klk1, Klk1l, Klk5, Klk7, RATKALA, RGK-7, RSKG-7
- Chromosome Location
-
Chr.1: 100131562 - 100135556 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_012593.1 |
NP_036725.1 |
2 - 3 |
205 |
118 |
View Full Product Details