The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_134385.2 | NP_599212.1 | 1 - 2 | 66 | 67 |
XM_006253905.1 | XP_006253967.1 | 1 - 2 | 60 | 67 | |
GenBank mRNA | AF239745.1 | - | 1 - 2 | 66 | 67 |
AF239747.1 | - | 1 - 2 | 66 | 67 | |
AF239748.1 | - | 1 - 2 | 66 | 67 | |
BC100138.1 | - | 1 - 2 | 59 | 67 | |
BC127474.1 | - | 1 - 2 | 57 | 67 |