The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145778.2 | NP_665721.1 | 9 - 10 | 1107 | 68 |
GenBank mRNA | AB015946.1 | - | 9 - 10 | 1012 | 68 |
BC070957.1 | - | 9 - 10 | 1107 | 68 |