The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_183325.2 | NP_899154.2 | 3 - 4 | 821 | 101 |
XM_006248360.2 | XP_006248422.1 | 3 - 4 | 1037 | 101 | |
GenBank mRNA | BC082065.1 | - | 3 - 4 | 821 | 101 |
M86341.1 | - | 2 - 3 | 676 | 101 |