The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013222.2 | NP_037354.2 | 2 - 3 | 512 | 119 |
GenBank mRNA | D30735.1 | - | 2 - 3 | 512 | 119 |