The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023100.1 | NP_075588.1 | 1 - 2 | 855 | 69 |
GenBank mRNA | AB038649.1 | - | 1 - 2 | 855 | 69 |
AF242873.1 | - | 1 - 2 | 825 | 69 | |
BC081796.1 | - | - | 882 | 69 |