The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019318.1 | NP_062191.1 | 1 - 1 | 1751 | 111 |
GenBank mRNA | U56242.1 | - | 1 - 1 | 1751 | 111 |