Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Morf4l1
Entrez Gene ID
300891
Gene Name
mortality factor 4 like 1
Gene Aliases
-
Chromosome Location
Chr.8: 97471523 - 97494974 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008766445.1 XP_008764667.1 2 - 3 196 117

View Full Product Details