Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Celf4
Entrez Gene ID
307540
Gene Name
CUGBP, Elav-like family member 4
Gene Aliases
Brunol4
Chromosome Location
Chr.18: 17689090 - 18079835 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107400.1 NP_001100870.1 9 - 10 1444 70

View Full Product Details