Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Spred3
Entrez Gene ID
308478
Gene Name
sprouty-related, EVH1 domain containing 3
Gene Aliases
RGD1311713
Chromosome Location
Chr.1: 88101213 - 88112683 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008759144.1 XP_008757366.1 2 - 3 253 128
XM_008759145.1 XP_008757367.1 2 - 3 200 128
XM_008759146.1 XP_008757368.1 2 - 3 322 128

View Full Product Details