The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270849.1 | NP_001257778.1 | 10 - 11 | 1457 | 60 |
NM_001270850.1 | NP_001257779.1 | 9 - 10 | 1235 | 60 | |
NM_012565.2 | NP_036697.1 | 9 - 10 | 1358 | 60 | |
XM_006251179.2 | XP_006251241.1 | 9 - 10 | 2203 | 60 | |
GenBank mRNA | J04218.1 | - | 9 - 10 | 1317 | 60 |
KJ026953.1 | - | 9 - 10 | 1250 | 60 | |
M25807.1 | - | 9 - 10 | 1235 | 60 | |
X53588.1 | - | 9 - 10 | 1341 | 60 | |
X53589.1 | - | 10 - 11 | 1457 | 60 |