Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tet3
Entrez Gene ID
680576
Gene Name
tet methylcytosine dioxygenase 3
Gene Aliases
-
Chromosome Location
Chr.4: 115060702 - 115160802 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001057850.5 XP_001057850.2 2 - 3 1892 90
XM_006236792.2 XP_006236854.1 2 - 3 1566 90
XM_008763094.1 XP_008761316.1 3 - 4 1190 90
XM_008763095.1 XP_008761317.1 3 - 4 3937 90

View Full Product Details