Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Map7d1
Entrez Gene ID
681287
Gene Name
MAP7 domain containing 1
Gene Aliases
Mtap7d1
Chromosome Location
Chr.5: 144250416 - 144274981 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109438.1 NP_001102908.1 6 - 7 1109 84
XM_006238901.2 XP_006238963.1 6 - 7 1089 84
XM_006238902.2 XP_006238964.1 6 - 7 1080 84

View Full Product Details