Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zbtb46
Entrez Gene ID
311718
Gene Name
zinc finger and BTB domain containing 46
Gene Aliases
Btbd4
Chromosome Location
Chr.3: 176888502 - 176958832 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107808.1 NP_001101278.1 2 - 3 1260 63
XM_008762510.1 XP_008760732.1 2 - 3 1265 63

View Full Product Details