Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Snx12
Entrez Gene ID
363478
Gene Name
sorting nexin 12
Gene Aliases
RGD1565585
Chromosome Location
Chr.X: 70989822 - 71127347 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108817.2 NP_001102287.2 2 - 3 319 78
XM_006257078.1 XP_006257140.1 2 - 3 388 78
XM_006257079.1 XP_006257141.1 2 - 3 388 78
GenBank mRNA BP500254.1 - 2 - 3 325 78

View Full Product Details