Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Notum
Entrez Gene ID
303743
Gene Name
notum pectinacetylesterase homolog (Drosophila)
Gene Aliases
RGD1307119
Chromosome Location
Chr.10: 109832977 - 109840047 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013975.2 NP_001013997.2 3 - 4 484 58
XM_006247887.2 XP_006247949.1 3 - 4 717 58
XM_006247888.2 XP_006247950.1 2 - 3 1054 58
GenBank mRNA DV215880.1 - 3 - 4 484 58
EX493402.1 - 2 - 3 288 58

View Full Product Details