Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tardbp
Entrez Gene ID
298648
Gene Name
TAR DNA binding protein
Gene Aliases
-
Chromosome Location
Chr.5: 165432089 - 165442112 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC083752.1 - 7 - 8 1189 127

View Full Product Details