Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Casc4
Entrez Gene ID
362204
Gene Name
cancer susceptibility candidate 4
Gene Aliases
RGD1305985
Chromosome Location
Chr.3: 113818649 - 113887550 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271283.1 NP_001258212.1 8 - 9 1310 81

View Full Product Details