Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hsd17b8
Entrez Gene ID
361802
Gene Name
hydroxysteroid (17-beta) dehydrogenase 8
Gene Aliases
Fabgl, Ke6, RING2
Chromosome Location
Chr.20: 3817212 - 3819323 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_212529.1 NP_997694.1 6 - 7 659 64
XM_006255964.2 XP_006256026.1 5 - 6 737 64
XM_006255965.2 XP_006256027.1 6 - 7 774 64
XM_006255966.2 XP_006256028.1 5 - 6 737 64
GenBank mRNA BC091424.1 - 3 - 4 971 64

View Full Product Details