The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033359.3 | NP_203523.2 | 3 - 4 | 724 | 74 |
GenBank mRNA | AF333245.1 | - | 3 - 4 | 315 | 74 |
AY066001.1 | - | 3 - 4 | 524 | 74 |