The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001025625.2 | NP_001020796.2 | 4 - 5 | 719 | 102 |
GenBank mRNA | CB326459.1 | - | 3 - 4 | 102 |