The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001106970.1 | NP_001100440.1 | 3 - 4 | 364 | 107 |
GenBank mRNA | BC168927.1 | - | 3 - 4 | 285 | 107 |