Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sap30l
Entrez Gene ID
360531
Gene Name
SAP30-like
Gene Aliases
RGD1309585
Chromosome Location
Chr.10: 43224151 - 43231864 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_340803.7 XP_340804.4 3 - 4 539 75

View Full Product Details