Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Map1b
Entrez Gene ID
29456
Gene Name
microtubule-associated protein 1B
Gene Aliases
Mtap1b
Chromosome Location
Chr.2: 29675391 - 29768750 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019217.1 NP_062090.1 2 - 3 451 95
GenBank mRNA BC158858.1 - 2 - 3 364 95
CB610210.1 - 2 - 3 451 95
U52950.1 - 2 - 3 347 95

View Full Product Details