Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sall3
Entrez Gene ID
364910
Gene Name
spalt-like transcription factor 3
Gene Aliases
-
Chromosome Location
Chr.18: 77572200 - 77591685 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108892.1 NP_001102362.1 1 - 2 3482 66
XM_006254985.2 XP_006255047.1 2 - 3 3597 66
XM_006254987.2 XP_006255049.1 3 - 4 3381 66

View Full Product Details