Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gp6
Entrez Gene ID
684555
Gene Name
glycoprotein 6 (platelet)
Gene Aliases
-
Chromosome Location
Chr.1: 73040713 - 73064639 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008759019.1 XP_008757241.1 1 - 2 321 63

View Full Product Details