Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pax3
Entrez Gene ID
114502
Gene Name
paired box 3
Gene Aliases
-
Chromosome Location
Chr.9: 84004004 - 84101230 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053710.1 NP_446162.1 1 - 2 471 114
XM_006245131.2 XP_006245193.1 1 - 2 528 114
XM_008767181.1 XP_008765403.1 1 - 2 529 114
GenBank mRNA AY169320.1 - 1 - 2 101 114

View Full Product Details