The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053710.1 | NP_446162.1 | 7 - 8 | 1556 | 59 |
XM_006245131.2 | XP_006245193.1 | 7 - 8 | 1610 | 59 | |
XM_008767181.1 | XP_008765403.1 | 8 - 9 | 1689 | 59 |