Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ptma
Entrez Gene ID
29222
Gene Name
prothymosin alpha
Gene Aliases
-
Chromosome Location
Chr.9: 93545396 - 93549436 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021740.1 NP_068508.1 1 - 2 193 59
GenBank mRNA BC061972.1 - 1 - 2 203 59
BC092569.1 - 1 - 2 213 59
FQ210626.1 - 1 - 2 224 59
FQ212611.1 - 1 - 2 224 59
FQ214221.1 - 1 - 2 218 59
FQ219686.1 - 1 - 2 224 59
FQ220677.1 - 1 - 2 225 59
FQ221801.1 - 1 - 2 224 59
FQ225103.1 - 1 - 2 228 59
FQ226946.1 - 1 - 2 224 59
FQ227835.1 - 1 - 2 224 59
FQ229930.1 - 1 - 2 224 59
FQ231540.1 - 1 - 2 224 59
FQ231794.1 - 1 - 2 230 59
FQ232039.1 - 1 - 2 228 59
FQ233285.1 - 1 - 2 230 59
M20035.1 - 1 - 2 220 59
M33024.1 - 1 - 2 220 59
M86564.1 - 1 - 2 193 59
X55326.1 - 1 - 2 193 59

Gene Symbol
LOC100359583
Entrez Gene ID
100359583
Gene Name
hypothetical protein LOC100359583
Gene Aliases
-
Chromosome Location
Chr.13: 89873071 - 89874031 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003751278.3 XP_003751326.1 1 - 1 230 59

View Full Product Details