Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Map2
Entrez Gene ID
25595
Gene Name
microtubule-associated protein 2
Gene Aliases
MAP2R, Mtap2
Chromosome Location
Chr.9: 73334908 - 73462972 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013066.1 NP_037198.1 10 - 11 5168 86
XM_008767203.1 XP_008765425.1 10 - 11 5348 86
XM_008767206.1 XP_008765428.1 10 - 11 5136 86
XM_008767208.1 XP_008765430.1 8 - 9 1154 86
GenBank mRNA X17682.1 - 7 - 8 1058 86
X51842.1 - 10 - 11 5140 86
X54100.1 - 10 - 11 5168 86

View Full Product Details