Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rpe
Entrez Gene ID
501157
Gene Name
ribulose-5-phosphate-3-epimerase
Gene Aliases
-
Chromosome Location
Chr.9: 73687682 - 73713643 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033903.1 NP_001029075.1 2 - 3 212 59
XM_006245107.2 XP_006245169.1 2 - 3 308 59
XM_006245109.2 XP_006245171.1 2 - 3 304 59
XM_008767290.1 XP_008765512.1 2 - 3 288 59
GenBank mRNA BC101869.1 - 2 - 3 212 59

View Full Product Details