The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033903.1 | NP_001029075.1 | 3 - 4 | 350 | 103 |
XM_006245104.2 | XP_006245166.1 | 4 - 5 | 539 | 103 | |
XM_006245106.2 | XP_006245168.1 | 4 - 5 | 502 | 103 | |
XM_006245109.2 | XP_006245171.1 | 3 - 4 | 442 | 103 | |
XM_008767290.1 | XP_008765512.1 | 3 - 4 | 426 | 103 | |
GenBank mRNA | BC101869.1 | - | 3 - 4 | 350 | 103 |